1,1'-(1,2-Ethanediyl)bis-1H-1,2,4-triazole ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
1,2-Di(1H-1,2,4-triazol-1-yl)ethane
CDX-E0066-M100
CDX-E0066-G001
102989-916EA
228.46
USD
102989-916
102989-918
1,1'-(1,2-Ethanediyl)bis-1H-1,2,4-triazole ≥97% (by HPLC)
1,1'-(1,2-Ethanediyl)bis-1H-1,2,4-triazole
Building block for synthesis.
Formula:
C₆H₈N₆ MW: 164.17 g/mol Storage Temperature: Refrigerator |
CAS Number:
116476-98-3 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...