Epoxidized linseed oil ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: ELO

CDX-E0038-G005 CDX-E0038-G025
102989-904EA 157.85 USD
102989-904 102989-906
Epoxidized linseed oil ≥98%
Epoxidized linseed oil

Useful for the determination of ELO in foods with HPLC-MS or GC-MS. ELO is currently employed as plasticizer and HCl scavenger in PVC products. Vegetable oils are widely used for chemical manipulation because of the high numbers of carbon-carbon double bonds. Through the epoxidation, an epoxide group (which is a more reactive group than a double bond) is added to the soybean oil, transforming it in a good hydrochloric acid scavenger and good plasticizer. It is also used as a reactive chemical intermediate due to its high oxirane content.

MW: g/mol
Storage Temperature: Freezer
MDL Number: MFCD00163559
CAS Number: 8016-11-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (CHN)
Appearance/Color White-yellow to brown oil.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR