2-Ethoxy-2-(2-naphthyl)acetonitrile 95
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-E0031-M050
CDX-E0031-M250
102989-896EA
222.92
USD
102989-896
102989-898
2-Ethoxy-2-(2-naphthyl)acetonitrile 95
2-Ethoxy-2-(2-naphthyl)acetonitrile
This is a new substrate for Cytochrom P450 suitable for fluorescence assays.
Formula:
C₁₄H₁₃NO MW: 211.26 g/mol Storage Temperature: Freezer |
CAS Number:
33224-80-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in acetonitrile. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...