Ethidium homodimer I ≥90%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: EthD-1, EtDi

CDX-E0012-M001 CDX-E0012-M005 CDX-E0012-M010
102989-880EA 258.92 USD
102989-880 102989-882 102989-884
Ethidium homodimer I ≥90%
Ethidium homodimer I

Staining dye for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. It does not cross intact cell membranes and can be used to test cell viability. Reagent for the fluorimetric detection of nucleic acids.

Formula: C₄₆H₅₀Cl₄N₈
MW: 856.75 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210948
CAS Number: 61926-22-5
UN: 3096
ADR: 8 (4.3) III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity ~90% (HPCE)
Appearance/Color Dark red solid.
Solubility Soluble in water, DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR