2-(1,1-Dimethylpropyl)-6-(diphenylphosphino)pyridine ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: ALPYPHOS

CDX-D0916-M500 CDX-D0916-GG25
102989-854EA 96.92 USD
102989-854 102989-856
2-(1,1-Dimethylpropyl)-6-(diphenylphosphino)pyridine ≥98% (by HPLC)
2-(1,1-Dimethylpropyl)-6-(diphenylphosphino)pyridine

Catalyst for regioselective synthesis of oligo-1,4-diols and anti-Markovnikov hydration.

Formula: C₂₂H₂₄NP
MW: 333.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10000873
CAS Number: 947315-18-6

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR