5-Hydroxyferulic acid ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3,4-Dihydroxy-5-methoxycinnamic acid

CDX-D0915-M010 CDX-D0915-M050 CDX-D0915-M100
102989-848EA 139.85 USD
102989-848 102989-850 102989-852
5-Hydroxyferulic acid ≥95% (by HPLC)
5-Hydroxyferulic acid

Hydroxycinnamic acid. Secondary plant metabolite of the phenylpropanoid pathway. It is a precursor in the biosynthesis of sinapic acid. Formed from ferulic acid by the action of the specific enzyme ferulate 5-hydroxylase (F5H).

Formula: C₁₀H₁₀O₅
MW: 210.18 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03002791
CAS Number: 1782-55-4

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR