DSP-1 ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3'-Methoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-6'-yl 2-fluoro-5-nitrobenzoate

CDX-D0391-M001 CDX-D0391-M005 CDX-D0391-M010
102989-808EA 157.85 USD
102989-808 102989-810 102989-812
DSP-1 ≥98%
DSP-1

Fluorescent probe for detection of H2S2 and hydrogen polysulfides in aqueous buffers.

Formula: C₂₈H₁₆FNO₈
Storage Temperature: Freezer
CAS Number: 1609395-29-0

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White to yellow floc.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR