(S)-O-Desmethylnaproxen ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
(S)-6-Hydroxy-alpha-methyl-2-naphthaleneacetic acid
CDX-D0390-M010
CDX-D0390-M050
102989-804EA
210.46
USD
102989-804
102989-806
(S)-O-Desmethylnaproxen ≥97% (by HPLC)
(S)-O-Desmethylnaproxen
Major naproxen demethylation metabolite. Naproxen demethylation is mediated primarily by CYP2C9. Can be used for assaying naproxen metabolites.
Formula:
C₁₃H₁₂O₃ MW: 216.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00869765 CAS Number: 52079-10-4 UN: 3077 ADR: 9,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...