N-Decanoyl-DL-homoserine lactone ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: C10-HSL

CDX-D0338-M010 CDX-D0338-M050
102989-796EA 123.23 USD
102989-796 102989-798
N-Decanoyl-DL-homoserine lactone ≥97%
N-Decanoyl-DL-homoserine lactone

N-Decanoyl-DL-homoserine lactone (N-C10-HSL) is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.

Formula: C₁₄H₂₅NO₃
MW: 255.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095526
CAS Number: 106983-36-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR