3,5-Difluoro-2,4-dihydroxybenzaldehyde ≥98% (by GC)
Supplier: ADIPOGEN CORP MS
CDX-D0330-M250
CDX-D0330-G001
102989-774EA
220.15
USD
102989-774
102989-776
3,5-Difluoro-2,4-dihydroxybenzaldehyde ≥98% (by GC)
3,5-Difluoro-2,4-dihydroxybenzaldehyde
Building block for synthesis. Used for the preparation of fluorescent probes.
Formula:
C₇H₄F₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD24675922 CAS Number: 209541-27-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...