Dinotefuran ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
1-Methyl-3-nitro-2-(tetrahydro-3-furanylmethyl)guanidine
CDX-D0309-M100
CDX-D0309-G001
102989-746EA
210.46
USD
102989-746
102989-748
Dinotefuran ≥97% (by HPLC)
Dinotefuran
A neonicotinoid insecticide. Compound can be used as analytical reference material.
Formula:
C₇H₁₄N₄O₃ MW: 202.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD06795001 CAS Number: 165252-70-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white powder/crystals. |
Solubility | Soluble in toluene. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...