cis-2-Dodecenoic acid ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: BDSF

CDX-D0293-M010 CDX-D0293-M250
102989-724EA 192.46 USD
102989-724 102989-726
cis-2-Dodecenoic acid ≥95% (by HPLC)
cis-2-Dodecenoic acid

Burkholderia cenocepacia produces a diffusible fatty acid signal molecule, cis-2-dodecenoic acid (BDSF), that has been implicated in interspecies and interkingdom communication.

Formula: C₁₂H₂₂O₂
MW: 198.3 g/mol
Storage Temperature: Freezer
CAS Number: 55928-65-9
UN: 3077
ADR: 9,

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Low melting solid.
Solubility Soluble in DMF, DMSO or chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR