DAR-4 ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Diaminorhodamine-4

CDX-D0276-M001 CDX-D0276-M005
102989-720EA 351.69 USD
102989-720 102989-722
DAR-4 ≥98%
DAR-4

Photo-stable fluorescent probe for the detection of nitric oxide (NO).

Formula: C₂₄H₂₄N₄O₃
Storage Temperature: Freezer
CAS Number: 339527-78-5

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color Dark violet powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR