Desacetylcolchicine d-tartrate ≥97% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
Trimethylcolchicinic acid
CDX-D0270-M005
CDX-D0270-M025
CDX-D0270-M100
102989-710EA
139.85
USD
102989-710
102989-712
102989-714
Desacetylcolchicine d-tartrate ≥97% (by NMR)
Desacetylcolchicine-d-tartrate
Anticancer colchicine analog. Microtubule polymerization inhibitor. Prevents cell division.
Formula:
C₂₀H₂₃NO₅·C₄H₆O₆ MW: 507.5 g/mol Storage Temperature: Refrigerator |
CAS Number:
49720-72-1 UN: 1544 ADR: 6.1,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Yellow to brown powder. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...