1,3-Dihydro-1-hydroxy-3,3-dimethyl-1,2-benziodoxole ≥99% (by HPLC)
Supplier: ADIPOGEN CORP MS
CDX-D0255-M100
CDX-D0255-G001
102989-706EA
210.46
USD
102989-706
102989-708
1,3-Dihydro-1-hydroxy-3,3-dimethyl-1,2-benziodoxole ≥99% (by HPLC)
1,3-Dihydro-1-hydroxy-3,3-dimethyl-1,2-benziodoxole
Heterocyclic lambda3-iodane. Building block for synthesis.
Formula:
C₉H₁₁IO₂ Storage Temperature: Freezer |
CAS Number:
69429-70-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Light yellow powder. |
Solubility | Soluble in toluene. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...