DAABD-AE ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 4-[2-(N,N-Dimethylamino)ethylaminosulphonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole

CDX-D0204-M050 CDX-D0204-M250
102989-660EA 333.69 USD
102989-660 102989-662
DAABD-AE ≥95% (by HPLC)
DAABD-AE

4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole (DAABD-AE) is new derivatization reagent for rapid and specific liquid chromatography tandem mass spectrometric quantification of pristanic acid, phytanic acid, and very long chain fatty acids (i.e. hexacosanoic, tetracosanoic, and docosanoic acids) in plasma.

Formula: C₁₂H₂₀N₆O₃S
MW: 328.39 g/mol
Storage Temperature: Freezer
CAS Number: 913253-56-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Orange powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR