7-Diethylamino-3-[N-(4-maleimidopropyl)carbamoyl]coumarin ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-D0199-M001 CDX-D0199-M010 CDX-D0199-M100
102989-650EA 38.77 USD
102989-650 102989-652 102989-654
7-Diethylamino-3-[N-(4-maleimidopropyl)carbamoyl]coumarin ≥97% (by HPLC)
7-Diethylamino-3-[N-(4-maleimidopropyl)carbamoyl]coumarin

Thiol-reactive probe for protein labelling.

Formula: C₂₁H₂₃N₃O₅
Storage Temperature: Freezer
CAS Number: 160291-54-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO, DMF or chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR