7-Diethylamino-3-[N-(4-maleimidoethyl)carbamoyl]coumarin ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-D0198-M001
CDX-D0198-M020
CDX-D0198-M250
102989-644EA
34.62
USD
102989-644
102989-646
102989-648
7-Diethylamino-3-[N-(4-maleimidoethyl)carbamoyl]coumarin ≥97% (by HPLC)
7-Diethylamino-3-[N-(4-maleimidoethyl)carbamoyl]coumarin
Thiol-reactive fluorescent probe for protein labeling. MDCC has been used for preparing specific probes for inorganic phosphate.
Formula:
C₂₀H₂₁N₃O₅ Storage Temperature: Freezer |
CAS Number:
156571-46-9 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Yellow powder. |
Solubility | Soluble in DMSO, DMF or chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...