2,3-Diaminopropane-1-thiol dihydrobromide ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 2,3-Diamino-1-propanethiol dihydrobromide

CDX-D0138-M025 CDX-D0138-M100 CDX-D0138-M250
102987-894EA 245.08 USD
102987-894 102987-896 102987-898
2,3-Diaminopropane-1-thiol dihydrobromide ≥95% (by HPLC)
2,3-Diaminopropane-1-thiol dihydrobromide

Building block for synthesis.

Formula: C₃H₁₀N₂S·2HBr
MW: 268 g/mol
Storage Temperature: Freezer
CAS Number: 70548-07-1

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in water, ethanol or ether.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR