DCIA ≥85% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 7-Diethylamino-3-[4'-(iodoacetamido)phenyl]-4-methylcoumarin

CDX-D0090-M010 CDX-D0090-M020 CDX-D0090-M100
102987-780EA 141.23 USD
102987-780 102987-782 102987-784
DCIA ≥85% (by HPLC)
DCIA

A label that can be used for preparing maleimide and iodoacetamide derivatives of 3-phenylcoumarin fluorophore. These derivatives can be self-quenching until they react with thiol groups and regain their strong fluorescence.

Formula: C₂₂H₂₃IN₂O₃
MW: 490.33 g/mol
Storage Temperature: Freezer
CAS Number: 76877-34-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 85% (HPLC)
Appearance/Color Dark yellow crystalline.
Solubility Soluble in DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR