DPM ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: N-[4-(5,6-Dimethoxy-N-phthalimidinyl)phenyl]maleimide

CDX-D0049-M025 CDX-D0049-M250
102987-724EA 318.46 USD
102987-724 102987-726
DPM ≥97%
DPM

Nonfluorescent N-substituted maleimides react selectively with thiol compounds to form fluorescent derivatives. Fluorescent intensity of the dimethoxyphthalimidyl group is higher than that of the nonsubsituted phthalimidyl group. The analytical sensitivity of N-[4-(5,6-Dimethoxy-N-phthalimidyl)phenyl]maleimide (DPM) is about 3-times higher than that of N-[4-(2-phthalimidyl)phenyl]maleimide (PPM). DPM can be used as a precolumn labeling reagent in HPLC. DPM was also used for the determination of physiologically important thiols in biological fluids or therapeutic drug monitoring.

Formula: C₂₀H₁₆N₂O₅
Storage Temperature: Freezer
CAS Number: 143503-03-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR