DACM ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: N-(7-Dimethylamino-4-methylcoumarin-3-yl) maleimide

CDX-D0047-M005 CDX-D0047-M025
102987-720EA 342 USD
102987-720 102987-722
DACM ≥98%
DACM

Excellent blue fluorescent thiol-reactive reagent (Ex/Em: 383/463nm), widely used for probing configurations of biomolecules such as proteins and create blue-fluorescent bioconjugates. The unreacted reagent is non-fluorescent and can also be used to quantitate free thiols. Shows superior fluorescence and water solubility properties than dansyl chloride.

Formula: C₁₆H₁₄N₂O₄
Storage Temperature: Freezer
CAS Number: 55145-14-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color Orange solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR