DAF-2T ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-D0044-M001 CDX-D0044-M005 CDX-D0044-M010
102987-706EA 368.31 USD
102987-706 102987-708 102987-710
DAF-2T ≥95% (by HPLC)
DAF-2T

Reference material for DAF-2 (CDX-D0084). Spectral data: lambdaex 495nm; lambdaem 515nm.

Formula: C₂₀H₁₁N₃O₅
MW: 373.32 g/mol
Storage Temperature: Freezer
CAS Number: 208850-35-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Yellow to orange solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR