7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM) ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 1H-Pyrrole-2,5-dione, 1-(4-(7-(diethylamino)-4-methyl-2-oxo-2H-1-benzopyran-3-yl)phenyl)-, CPM

CDX-D0042-M050 CDX-D0042-M250
102987-702EA 342 USD
102987-702 102987-704
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM) ≥95%
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM)

Widely used blue fluorescent thiol-reactive dye (Ex/Em: 384/470nm). This maleimide derivative of coumarin is essentially non-fluorescent until it reacts with thiols, making it possible to quantify thiols without a separation step. Good energy acceptor from tryptophan and a good energy donor to fluorescein. Used to monitor release of thiols and to distinguish proliferating cancer cells by nucleolar protein staining.

Formula: C₂₄H₂₂N₂O₄
MW: 402.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077334
CAS Number: 76877-33-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >95% (HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO, DMF, acetonitrile or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR