7-(Diethylamino)coumarin-3-carbohydrazide ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 7-(Diethylamino)-2-oxo-2H-chromene-3-carbohydrazide, 7-Diethylaminocoumarin-3-carbohydrazide

CDX-D0035-M050 CDX-D0035-M250
102987-684EA 333.69 USD
102987-684 102987-686
7-(Diethylamino)coumarin-3-carbohydrazide ≥95% (by HPCE)
7-(Diethylamino)coumarin-3-carbohydrazide

Derivatizing agent for carboxylic acid detection. Has also been used for fluorescence detection of monoterpene alcohols.

Formula: C₁₄H₁₇N₃O₃
MW: 275.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467583
CAS Number: 100343-98-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in methanol, DMF, acetonitrile or chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR