6,7-Dimethoxy-4-coumarinylacetic acid ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS

CDX-D0034-M100 CDX-D0034-G001
102987-680EA 276.92 USD
102987-680 102987-682
6,7-Dimethoxy-4-coumarinylacetic acid ≥98% (by TLC)
6,7-Dimethoxy-4-coumarinylacetic acid

6,7-Dimethoxy-4-coumarinyl acetic is a fluorescent molecule which may be used in fluorescent applications that use coumarinyl acetic acids.

Formula: C₁₃H₁₂O₆
Storage Temperature: Refrigerator
CAS Number: 88404-26-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color Light tan solid.
Solubility Soluble in DMSO or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR