6,7-Dimethoxy-4-(trifluoromethyl) coumarin ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-D0033-M100 CDX-D0033-G001
102987-676EA 296.31 USD
102987-676 102987-678
6,7-Dimethoxy-4-(trifluoromethyl) coumarin ≥95% (by HPLC)
6,7-Dimethoxy-4-(trifluoromethyl) coumarin

Fluorescent labelling.

Formula: C₁₂H₉F₃O₄
Storage Temperature: Refrigerator
CAS Number: 151625-32-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR