6,7-Dihydroxy-4-coumarinylacetic acid ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-D0029-M100 CDX-D0029-G001
102987-668EA 258.92 USD
102987-668 102987-670
6,7-Dihydroxy-4-coumarinylacetic acid ≥95% (by HPLC)
6,7-Dihydroxy-4-coumarinylacetic acid

Fluorescent label.

Formula: C₁₁H₈O₆
Storage Temperature: Refrigerator
CAS Number: 88404-14-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Pink solid.
Solubility Soluble in DMSO or water (pH 6).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR