6,7-Diethoxy-4-methylcoumarin ≥95%

Supplier: ADIPOGEN CORP MS

CDX-D0028-M100 CDX-D0028-G001
102987-664EA 278.31 USD
102987-664 102987-666
6,7-Diethoxy-4-methylcoumarin ≥95%
6,7-Diethoxy-4-methylcoumarin

Fluorogenic dealkylase substrate for the measurement of the cytochrome-mediated monooxygenase or "mixed function" oxidase system.

Formula: C₁₄H₁₆O₄
MW: 248.27 g/mol
Storage Temperature: Refrigerator
CAS Number: 314744-06-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMF, acetonitrile, alcohols or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR