6,7-Diethoxy-4-(trifluoromethyl) coumarin ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-D0027-M100 CDX-D0027-G001
102987-660EA 278.31 USD
102987-660 102987-662
6,7-Diethoxy-4-(trifluoromethyl) coumarin ≥95% (by HPLC)
6,7-Diethoxy-4-(trifluoromethyl)coumarin

Fluorescent labelling.

Formula: C₁₄H₁₃F₃O₄
MW: 302.25 g/mol
Storage Temperature: Refrigerator
CAS Number: 351002-66-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in chloroform or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR