4-(N,N-Dimethylaminomethyl)-7-propoxycoumarin ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-D0020-M050 CDX-D0020-M500
102989-630EA 275.54 USD
102989-630 102989-632
4-(N,N-Dimethylaminomethyl)-7-propoxycoumarin ≥97% (by HPLC)
4-(N,N-Dimethylaminomethyl)-7-propoxycoumarin

Fluorescent labelling.

Formula: C₁₅H₁₉NO₃
Storage Temperature: Freezer
CAS Number: 351194-17-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR