4-Di-2-ASP ≥97% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4-(4-Diethylaminostyryl)-1-methylpyridinium iodide

CDX-D0012-G001 CDX-D0012-G005
102989-614EA 211.85 USD
102989-614 102989-616
4-Di-2-ASP ≥97% (by NMR)
4-Di-2-ASP

Some cationic mitochondrial dyes such as 4-Di-1-ASP and 4-Di-2-ASP stain presynaptic nerve terminals independent of neuronal activity. The photostable 4-Di-2-ASP dye, which is nontoxic to cells, has been employed to stain living nerve terminals in rabbit corneal epithelium, in rat epidermis and at mouse, snake and frog neuromuscular junctions, as well as to visualize the innervation of the human choroid and whole mounts of the gastrointestinal tract. Methods for using 4-Di-2-ASP to image neuronal cells in live animals have been described (Ex/Em = 488/607 nm).

Formula: C₁₈H₂₃IN₂
MW: 394.29 g/mol
Storage Temperature: Refrigerator
CAS Number: 105802-46-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMSO, DMF or chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR