DIB-Cl ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
4-(4,5-Diphenyl-imidazol-2-yl)benzoyl-chloride
4-(4,5-diphenyl-1H-imidazol-2-yl)benzoyl chloride (DIB-Cl) as reagent for amine labelling was used successfully for derivatization and fluorescence detection of enantiomers of methamphetamin and p-hydroxymethamphetamin, in urine samples. It has also been used for 3,4-Methylendioxy-N-methylamphetamin in blood, morpholine and orther compounds. DIB-Cl as a label was used on fluorescent derivatization some hydroxyl group containing endocrine disruptors, including bisphenol A (BPA), bisphenol B (BPB), bisphenol E (BPE), bisphenol F (BPF) and 4-nonylphenol (4-NP). DIB-Cl showed superior fluorescence intensity (FI) to other commercially available reagents, excellent stability in solution and high reactivity with primary and secondary amines under mild conditions.
Formula:
C₂₂H₁₅ClN₂O MW: 358.82 g/mol Storage Temperature: Freezer |
CAS Number:
162756-62-9 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in acetonitrile. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...