1-(4'-Carboxybutyl)-3,7-dimethylxanthine ≥99%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 2,3,6,7-Tetrahydro-3,7-dimethyl-2,6-dioxo-1H-purine-1-pentanoic acid DB-dmx

CDX-C0312-M010 CDX-C0312-M025
102989-592EA 157.85 USD
102989-592 102989-594
1-(4'-Carboxybutyl)-3,7-dimethylxanthine ≥99%
1-(4'-Carboxybutyl)-3,7-dimethylxanthine

Pentoxifylline (PTX) metabolite. Can be used as reference material. Showed platelet aggregation in whole blood in vitro. Shown to be more effective than PTX in protecting WC/1 murine fibrosarcoma cells from TNF-alpha cytotoxicity and in inhibiting neutrophil superoxide anion production, degranulation (lactoferrin release) and surface expression of the beta-2 integrin CD11b/CD18.

Formula: C₁₂H₁₆N₄O₄
Storage Temperature: Freezer
CAS Number: 38975-44-9

Order Now

Specification Test Results

Source/Host Synthetic
Purity 99% (HPLC)
Appearance/Color White powder.
Solubility Soluble in ethyl acetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR