1-(3'-Carboxypropyl)-3,7-dimethylxanthine ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 4-(3,7-Dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)butanoic acid

CDX-C0311-M010 CDX-C0311-M050
102989-588EA 88.62 USD
102989-588 102989-590
1-(3'-Carboxypropyl)-3,7-dimethylxanthine ≥98%
1-(3'-Carboxypropyl)-3,7-dimethylxanthine

Major metabolite of pentoxifylline. Most effective metabolite at reducing oxygen radical production by polymorphonuclear leukocytes (PMN).

Formula: C₁₁H₁₄N₄O₄
Storage Temperature: Freezer
CAS Number: 6493-07-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR