Copper(II) tartrate hydrate ≥98% (by titrimetric analysis)

Supplier: ADIPOGEN CORP MS

Synonyms: Cupric tartrate hydrate, Tartaric acid cupric salt hydrate

CDX-C0241-G025 CDX-C0241-G100
102989-572EA 193.85 USD
102989-572 102989-574
Copper(II) tartrate hydrate ≥98% (by titrimetric analysis)
Copper(II) tartrate hydrate

Used as a chiral selector in electrophoresis. Substance for proteomics research.

Formula: C₄H₄CuO₆·xH₂O
MW: 211.62 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150650
CAS Number: 946843-80-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >98% (Titration)
Appearance/Color Blue crystalline powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR