1,4-Cubanedicarboxylic acid ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: Pentacyclo[4.2.0.02,5.03,8.04,7]octane-1,4-dicarboxylic acid

CDX-C0236-M250 CDX-C0236-G001
102989-560EA 279.69 USD
102989-560 102989-562
1,4-Cubanedicarboxylic acid ≥97%
1,4-Cubanedicarboxylic acid

Metal-organic frameworks (MOFs).

Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458496
CAS Number: 32846-66-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR