Cyflufenamid ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
(Z)-N-[alpha-(Cyclopropylmethoxyimino)-2,3-difluoro-6-(trifluoromethyl)benzyl]-2-phenylacetamide
CDX-C0230-M025
CDX-C0230-M100
CDX-C0230-M500
102989-550EA
105.23
USD
102989-550
102989-552
102989-554
Cyflufenamid ≥98% (by HPLC)
Cyflufenamid
Amidoxime fungicide used in agroculture. Active against powdery mildew of various crops in pre and post infection treatment. Has a long lasting translaminar and vapor phase activity. The mode of action is not know and it is considered different from that of other existing fungicides. It has favorable toxicological, ecotoxicological and environmental profiles. Compound can be used as analytical reference material.
Formula:
C₂₀H₁₇F₅N₂O₂ MW: 412.36 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16652594 CAS Number: 180409-60-3 UN: 3077 ADR: 9,III |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in methanol, ethanol or hexane. Sparingly soluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...