6-Chloro-2-methoxy-9-phenoxyacridine ≥98%
Supplier: ADIPOGEN CORP MS
|
Danger
|
CDX-C0178-G001
CDX-C0178-G002
CDX-C0178-G010
102989-524EA
170.31
USD
102989-524
102989-526
102989-528
6-Chloro-2-methoxy-9-phenoxyacridine ≥98%
6-Chloro-2-methoxy-9-phenoxyacridine
Acridine. Halogenated heterocycle used as building block.
Formula:
C₂₀H₁₄ClNO₂ Storage Temperature: Refrigerator |
CAS Number:
7478-26-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Yellow crystalline solid. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...