2-Cyclopropyl-propylamine -p-toluenesulfonate salt ≥98% (by elemental analysis)

Supplier: ADIPOGEN CORP MS

CDX-C0145-G002 CDX-C0145-G010
102989-500EA 258.92 USD
102989-500 102989-502
2-Cyclopropyl-propylamine -p-toluenesulfonate salt ≥98% (by elemental analysis)
2-Cyclopropylpropylamine-p-toluenesulfonate salt

Building block for synthesis.

Formula: C₆H₁₃N·C₇H₈O₃S
MW: 271.38 g/mol
Storage Temperature: Refrigerator
CAS Number: 172947-14-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (Elemental Analysis)
Appearance/Color Solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR