Carbonate ionophore VII ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
N,N-Dioctyl-3alpha,12alpha-bis(4-trifluoroacetylbenzoyloxy)-5beta-cholan-24-amide
CDX-C0131-M050
CDX-C0131-M250
102989-484EA
368.31
USD
102989-484
102989-486
Carbonate ionophore VII ≥95% (by HPLC)
Carbonate ionophore VII
Analytical/Chromatography reagent. Ion sensor compound. Ionophore potentiometric for ISE.
Formula:
C₅₈H₇₉F₆NO₇ MW: 1016.24 g/mol Storage Temperature: Refrigerator |
CAS Number:
222310-82-9 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...