Copper(II) ionophore IV ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: N,N,N′,N′-Tetracyclohexyl-2,2′-thiodiacetamide

CDX-C0130-M050 CDX-C0130-M250
102989-480EA 210.46 USD
102989-480 102989-482
Copper(II) ionophore IV ≥95%
Copper(II) ionophore IV

Analytical/Chromatography reagent. Ion sensor compound. Ionophore potentiometric for ISE.

Formula: C₂₈H₄₈N₂O₂S
MW: 476.76 g/mol
Storage Temperature: Refrigerator
CAS Number: 849629-03-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR