6-CFDA N-succinimidyl ester ≥90% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
6-Carboxyfluorescein diacetate N-succinimidyl ester
CDX-C0095-M005
CDX-C0095-M025
CDX-C0095-M100
102989-468EA
171.69
USD
102989-468
102989-470
102989-472
6-CFDA N-succinimidyl ester ≥90% (by HPLC)
6-CFDA-N-succinimidyl ester
6-CFDA is membrane-permeant and thus can be loaded into cells via incubation. Once inside the cells, 6-CFDA is hydrolyzed by intracellular esterases to 6-carboxyfluorescein. Reagent for continuously determining the intracellular pH in bacteria by fluorometry. Labeling human intervertebral disk cells in vitro for fluorescence microscopy.
Formula:
C₂₉H₁₉NO₁₁ MW: 557.46 g/mol Storage Temperature: Freezer |
CAS Number:
150206-15-8 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Light yellow powder. |
Solubility | Soluble in DMSO or DMF. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...