5-CFDA N-succinimidyl ester ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
5-CFDA N-succinimidyl ester, 5-Carboxyfluorescein diacetate N-succinimidyl ester
CDX-C0094-M005
CDX-C0094-M025
CDX-C0094-M100
102989-462EA
170.31
USD
102989-462
102989-464
102989-466
5-CFDA N-succinimidyl ester ≥95% (by HPLC)
5-CFDA-N-succinimidyl ester
5-CFDA is membrane-permeant and can be loaded into cells via incubation. Once inside the cells, 5-CFDA is hydrolyzed by intracellular esterases to 5-carboxyfluorescein. It has been used for labeling human intervertebral disk cells in vitro for fluorescence microscopy. Continuous determination of the intracellular pH in bacteria by fluorometry.
Formula:
C₂₉H₁₉NO₁₁ MW: 557.46 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00037466 CAS Number: 150206-05-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Powder. |
Solubility | Soluble in DMSO or DMF. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...