6-TAMRA ≥90% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate

CDX-C0056-M025 CDX-C0056-M250
102987-642EA 322.62 USD
102987-642 102987-644
6-TAMRA ≥90% (by HPLC)
6-TAMRA

Free acid form of 6-carboxytetramethylrhodamine single isomer. The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.

Formula: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01073638
CAS Number: 91809-67-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Black solid.
Solubility Soluble in methanol, DMSO, DMF or aqueous buffers (pH 6.5).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR