5-CFDA ethanedioic-S-phenylmethyl ester ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
CDX-C0051-M010
CDX-C0051-M050
102987-634EA
263.08
USD
102987-634
102987-636
5-CFDA ethanedioic-S-phenylmethyl ester ≥98% (by HPLC)
5-CFDA ethanedioic-S-phenylmethyl ester
Cell permeable fluorescent probe. Used for site-specific labeling of N-terminal cysteine proteins with the small molecule probes.
Formula:
C₃₄H₂₅NO₉S Storage Temperature: Freezer |
CAS Number:
690958-19-1 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...