5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥85% (by HPCE)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Carboxy-DCFDA, 5-(6)-Carboxydichlorofluorescein diacetate, 5(6)-CDCFDA, CDCFDA
CDX-C0042-M100
CDX-C0042-G001
102987-614EA
168.92
USD
102987-614
102987-616
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate ≥85% (by HPCE)
5(6)-Carboxy-2′,7′- dichlorofluorescein diacetate
Intracellular esterase substrate with low sensitivity of fluorescence to intracellar pH.
Formula:
C₂₅H₁₄Cl₂O₉ MW: 529.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08459585 CAS Number: 127770-45-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity | >85% (HPCE) |
Appearance/Color | White to off-white powder. |
Solubility | Soluble in DMSO, DMF or methanol. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...