3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: PacBlue succinimidyl ester, NHS Pacific Blue succinimidyl ester

CDX-C0036-M025 CDX-C0036-M100
102987-590EA 193.85 USD
102987-590 102987-592
3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥98% (by HPLC)
3-Carboxy-6,8-difluoro-7-hydroxycoumarin

This amine-reactive reagent can be used to create blue-fluorescent bioconjugates with excitation/emission maxima ~410/455 nm that are excitable by the 405 nm spectral line of the blue diode (violet) laser. Used in highly sensitive analyses of amines, amino acids and dipeptides. Amine-reactive reagents react with non-protonated aliphatic amine groups, including the amine terminus of proteins and the є-amino group of lysines.

Formula: C₁₄H₇F₂NO₇
MW: 339.2 g/mol
Storage Temperature: Freezer
CAS Number: 215868-33-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Yellow powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR