5(6)-ROX N-succinimidyl ester ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-C0013-M010 CDX-C0013-M025 CDX-C0013-M500
102987-544EA 166.15 USD
102987-544 102987-546 102987-548
5(6)-ROX N-succinimidyl ester ≥95% (by HPCE)
5(6)-ROX N-succinimidyl ester

5-(and-6)-Carboxy-X-rhodamine, succinimidyl ester is an amine-reactive long wavelength rhodamine dye. Derivatives made with the amine-reactive carboxy-X-rhodamine (ROX), succinimidyl ester are widely used for oligonucleotide labeling and automated DNA sequencing applications. Conjugates of this dye have longer-wavelength spectra than the spectra of Lissamine rhodamine B conjugates, but somewhat shorter-wavelength spectra than those of Texas Red® conjugates.

Formula: C₃₇H₃₃N₃O₇
Storage Temperature: Freezer
CAS Number: 114616-32-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Purple solid.
Solubility Soluble in DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR