Cholesteryl coumarin-3-carboxylate ≥96% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 3-CCA-Cl

CDX-C0006-M050 CDX-C0006-M250
102987-522EA 318.46 USD
102987-522 102987-524
Cholesteryl coumarin-3-carboxylate ≥96% (by HPLC)
Cholesteryl coumarin-3-carboxylate

Free coumarin-3-carboxylic acid shows a stronger fluorescence (Em. max: 419 nm (Exc.: 366 nm in ethanol).

Formula: C₃₇H₅₀O₄
MW: 558.79 g/mol
Storage Temperature: Refrigerator
CAS Number: 196091-78-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 96% (HPLC)
Appearance/Color Solid.
Solubility Soluble in dioxane.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR